4-Amino-3-chlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Amino-3-chlorobenzeneboronic acid, pinacol ester, 4-Amino-3-chlorophenylboronic Acid Pinacol Ester

PN-8923-5G PN-8923-1G
COBBPN-8923-5GEA 817 EUR
COBBPN-8923-5G COBBPN-8923-1G
4-Amino-3-chlorphenylboronsäurepinakolester
4-Amino-3-chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Molecular Weight: 253,54 g/mol
MDL: MFCD09998359
CAS-Nummer: 721960-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR