4-Chlor-3-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-3-nitrobenzeneboronic acid pinacol ester, 4-Chloro-3-nitrophenylboronic acid, pinacol ester, 2-(4-Chloro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2037-5G PN-2037-1G
COBBPN-2037-5GEA 725 EUR
COBBPN-2037-5G COBBPN-2037-1G
4-Chlor-3-nitrophenylboronsäurepinakolester
4-Chlor-3-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₅BClNO₄
Molecular Weight: 283.52 g/mol
MDL: MFCD08235086
CAS-Nummer: 913836-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR