2,4-Difluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(2,4-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2274-1G PN-2274-5G
COBBPN-2274-1GEA 61.9 EUR
COBBPN-2274-1G COBBPN-2274-5G
2,4-Difluorphenylboronsäurepinakolester
2,4-Difluorphenylboronsäurepinakolester
Formel: C₁₂H₁₅BF₂O₂
Molecular Weight: 240,06 g/mol
MDL: MFCD05663880
CAS-Nummer: 288101-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR