2,6-Difluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Difluorobenzeneboronic acid pinacol ester, 2,6-Difluorophenylboronic acid, pinacol ester, 2-(2,6-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2659-1G PN-2659-5G
COBBPN-2659-1GEA 0 EUR
COBBPN-2659-1G COBBPN-2659-5G
2,6-Difluorphenylboronsäurepinakolester
2,6-Difluorphenylboronsäurepinakolester
Formel: C₁₂H₁₅BF₂O₂
Molecular Weight: 240.06 g/mol
MDL: MFCD09801043
CAS-Nummer: 863868-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR