3,5-Difluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1,3-Difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-(3,5-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Difluorophenylboronic acid Pinacol ester

PN-2608-5G PN-2608-1G
COBBPN-2608-5GEA 352 EUR
COBBPN-2608-5G COBBPN-2608-1G
3,5-Difluorphenylboronsäurepinakolester
3,5-Difluorphenylboronsäurepinakolester
Formel: C₁₂H₁₅BF₂O₂
Molecular Weight: 240,06 g/mol
MDL: MFCD11504963
CAS-Nummer: 863868-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR