3-Cyan-2-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Cyano-2-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

PN-5008-5G PN-5008-1G
COBBPN-5008-5GEA 1180 EUR
COBBPN-5008-5G COBBPN-5008-1G
3-Cyan-2-fluorphenylboronsäurepinakolester
3-Cyan-2-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₅BFNO₂
Molecular Weight: 247,08 g/mol
MDL: MFCD11506358
CAS-Nummer: 1218791-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR