3-Cyan-4-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 2-(3-Cyano-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Cyano-4-fluorophenylboronic acid Pinacol ester, 3-Cyano-4-fluorophenylboronic acid, pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

PN-2327-5G PN-2327-1G
COBBPN-2327-5GEA 236 EUR
COBBPN-2327-5G COBBPN-2327-1G
3-Cyan-4-fluorphenylboronsäurepinakolester
3-Cyan-4-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₅BFNO₂
Molecular Weight: 247,08 g/mol
MDL: MFCD06795681
CAS-Nummer: 775351-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR