4-Amino-2,5-difluorphenylboronsäurepinakolester 96%
Lieferant: Thermo Fisher Scientific
Synonyme:
4-Amino-2,5-difluorophenylboronic acid pinacol ester, 2,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylamine
H52356.MD
H52356.03
H52356.MDEA
185
EUR
H52356.MD
H52356.03
4-Amino-2,5-difluorphenylboronsäurepinakolester 96%
4-Amino-2,5-difluorphenylboronsäurepinakolester
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
Purity: 96%
Molecular Formula: C12H16BF2NO2
Formula Weight: 255.07
MDL No.: MFCD09743045
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...