4-Amino-3-fluorphenylboronsäurepinakolester 96%

Lieferant: Thermo Scientific

Synonyme: 4-Amino-3-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Amino-3-fluorophenylboronic acid, pinacol ester

H52349.MD H52349.03
H52349.MDEA 29.2 EUR
H52349.MD H52349.03
4-Amino-3-fluorphenylboronsäurepinakolester 96%
4-Amino-3-fluorphenylboronsäurepinakolester

Purity: 96% CAS No.: 819058-34-9 Molecular Formula: C12H17BFNO2 Formula Weight: 237.08 MDL No.: MFCD09033884

Formel: C₁₂H₁₇BFNO₂
Molecular Weight: 237,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09033884
CAS-Nummer: 819058-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR