4-Amino-3-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Amino-3-nitrophenylboronic Acid Pinacol Ester, 4-Amino-3-nitrophenylboronic acid, pinacol ester

PN-8679-5G PN-8679-1G
COBBPN-8679-5GEA 375 EUR
COBBPN-8679-5G COBBPN-8679-1G
4-Amino-3-nitrophenylboronsäurepinakolester
4-Amino-3-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₇BN₂O₄
Molecular Weight: 264,09 g/mol
MDL: MFCD06795680
CAS-Nummer: 833486-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR