4-Amino-3-nitrophenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Amino-3-nitrophenylboronic Acid Pinacol Ester, 4-Amino-3-nitrophenylboronic acid, pinacol ester
M-Clarity™ Qualitätsstufe = MQ100
651613-1G
651613-5G
SIAL651613-1GEA
101
EUR
SIAL651613-1G
SIAL651613-5G
4-Amino-3-nitrophenylboronsäurepinakolester, Sigma-Aldrich®
4-Amino-3-nitrophenylboronsäurepinakolester
Formel:
C₁₂H₁₇BN₂O₄ Molecular Weight: 264,09 g/mol |
MDL:
MFCD06795680 CAS-Nummer: 833486-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...