2-Fluor-3-nitrophenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(2-Fluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2258-1G
PN-2258-250M
COBBPN-2258-1GEA
286
EUR
COBBPN-2258-1G
COBBPN-2258-250M
2-Fluor-3-nitrophenylboronsäurepinakolester
2-Fluor-3-nitrophenylboronsäurepinakolester
Formel:
C₁₂H₁₅BFNO₄ Molecular Weight: 267,06 g/mol |
MDL:
MFCD15143593 CAS-Nummer: 1189042-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...