2-Fluor-4-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-4-nitrobenzeneboronic acid pinacol ester, 2-(2-Fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5185-1G PN-5185-250M
COBBPN-5185-1GEA 1200 EUR
COBBPN-5185-1G COBBPN-5185-250M
2-Fluor-4-nitrophenylboronsäurepinakolester
2-Fluor-4-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₅BFNO₄
Molecular Weight: 267,06 g/mol
MDL: MFCD09264075
CAS-Nummer: 1073353-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR