2-Fluor-5-nitrophenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-5-nitrophenylboronic acid, pinacol ester, 2-(2-Fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-nitrobenzeneboronic acid pinacol ester
PN-5186-1G
PN-5186-5G
COBBPN-5186-1GEA
50.8
EUR
COBBPN-5186-1G
COBBPN-5186-5G
2-Fluor-5-nitrophenylboronsäurepinakolester
2-Fluor-5-nitrophenylboronsäurepinakolester
Formel:
C₁₂H₁₅BFNO₄ Molecular Weight: 267,06 g/mol |
MDL:
MFCD06409955 CAS-Nummer: 425378-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...