3-Fluor-4-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-3-nitrobenzeneboronic acid pinacol ester, 2-(3-Fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Fluoro-4-nitrobenzeneboronic acid pinacol ester

PN-5491-1G PN-5491-250M
COBBPN-5491-1GEA 812 EUR
COBBPN-5491-1G COBBPN-5491-250M
3-Fluor-4-nitrophenylboronsäurepinakolester
3-Fluor-4-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₅BFNO₄
Molecular Weight: 267,06 g/mol
MDL: MFCD12026073
CAS-Nummer: 939968-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR