3,4-Difluor-5-methoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(3,4-Difluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,4-Difluoro-5-methoxyphenylboronic acid, pinacol ester
PN-9054-1G
PN-9054-250M
COBBPN-9054-1GEA
667
EUR
COBBPN-9054-1G
COBBPN-9054-250M
3,4-Difluor-5-methoxyphenylboronsäurepinakolester
3,4-Difluor-5-methoxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BF₂O₃ Molecular Weight: 270,08 g/mol |
MDL:
MFCD15143617 CAS-Nummer: 754226-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...