(±)-2-Phenyl-3-butin-2-ol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Phenylbut-3-yn-2-ol
M-Clarity™ Qualitätsstufe = MQ200
212997-100G
212997-25G
212997-5G
SIAL212997-100GEA
268
EUR
SIAL212997-100G
SIAL212997-25G
SIAL212997-5G
(±)-2-Phenyl-3-butin-2-ol, Sigma-Aldrich®
(±)-2-Phenyl-3-butin-2-ol
Formel:
C₁₀H₁₀O Molecular Weight: 146,19 g/mol |
MDL:
MFCD00004454 CAS-Nummer: 127-66-2 EINECS: 204-855-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...