Thiomorpholin

Lieferant: FLUOROCHEM

019453-5G 019453-25G 019453-100G
FLUO019453-5GEA 39.9 EUR
FLUO019453-5G FLUO019453-25G FLUO019453-100G
Thiomorpholin
Thiomorpholin
Formel: C₄H₉NS
MDL: MFCD00005974
CAS-Nummer: 123-90-0
EINECS: 204-660-2
UN: 3267
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR