Thiomorpholin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
196274-25G
196274-5G
SIAL196274-25GEA
298
EUR
SIAL196274-25G
SIAL196274-5G
Thiomorpholin, Sigma-Aldrich®
Thiomorpholin
Formel:
C₄H₉NS Molecular Weight: 103,19 g/mol |
MDL:
MFCD00005974 CAS-Nummer: 123-90-0 EINECS: 204-660-2 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...