2-Amino-4-methylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Amino-4-methylphenylboronic acid, pinacol ester

PN-8678-5G PN-8678-1G
COBBPN-8678-5GEA 1520 EUR
COBBPN-8678-5G COBBPN-8678-1G
2-Amino-4-methylphenylboronsäurepinakolester
2-Amino-4-methylphenylboronsäurepinakolester
Formel: C₁₃H₂₀BNO₂
Molecular Weight: 233,12 g/mol
MDL: MFCD06795679
CAS-Nummer: 863578-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR