3-Amino-2-methylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PN-9127-1G PN-9127-5G
COBBPN-9127-1GEA 262 EUR
COBBPN-9127-1G COBBPN-9127-5G
3-Amino-2-methylphenylboronsäurepinakolester
3-Amino-2-methylphenylboronsäurepinakolester
Formel: C₁₃H₂₀BNO₂
Molecular Weight: 233.12 g/mol
MDL: MFCD11054038
CAS-Nummer: 882678-96-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR