4-Amino-2-methylphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-Amino-2-methylphenylboronic acid, pinacol ester, 3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
PN-8904-1G
PN-8904-250M
COBBPN-8904-1GEA
548
EUR
COBBPN-8904-1G
COBBPN-8904-250M
4-Amino-2-methylphenylboronsäurepinakolester
4-Amino-2-methylphenylboronsäurepinakolester
Formel:
C₁₃H₂₀BNO₂ Molecular Weight: 233,12 g/mol |
MDL:
MFCD08458198 CAS-Nummer: 631911-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...