3-Amino-4-methylphenylboronsäurepinakolester Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Amino-4-methylphenylboronic acid, pinacol ester, HCl

PN-2458-5G PN-2458-1G
COBBPN-2458-5GEA 1060 EUR
COBBPN-2458-5G COBBPN-2458-1G
3-Amino-4-methylphenylboronsäurepinakolester Hydrochlorid
3-Amino-4-methylphenylboronsäurepinakolester Hydrochlorid
Formel: C₁₃H₂₁BClNO₂
Molecular Weight: 269,58 g/mol
MDL: MFCD06659921
CAS-Nummer: 850567-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR