2-Methyl-3-nitrophenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4,4,5,5-Tetramethyl-2-(2-methyl-3-nitrophenyl)-1,3,2-dioxaborolane, 2-Methyl-3-nitrophenylboronic acid, pinacol ester
PN-2474-5G
PN-2474-1G
COBBPN-2474-5GEA
1010
EUR
COBBPN-2474-5G
COBBPN-2474-1G
2-Methyl-3-nitrophenylboronsäurepinakolester
2-Methyl-3-nitrophenylboronsäurepinakolester
Formel:
C₁₃H₁₈BNO₄ Molecular Weight: 263.1 g/mol |
MDL:
MFCD10699700 CAS-Nummer: 910235-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...