2-Methyl-5-nitrophenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methyl-5-nitrophenylboronic acid, pinacol ester, 2-Methyl-5-nitrophenylboronic Acid Pinacol Ester
PN-2473-5G
PN-2473-1G
COBBPN-2473-5GEA
681
EUR
COBBPN-2473-5G
COBBPN-2473-1G
2-Methyl-5-nitrophenylboronsäurepinakolester
2-Methyl-5-nitrophenylboronsäurepinakolester
Formel:
C₁₃H₁₈BNO₄ Molecular Weight: 263,1 g/mol |
MDL:
MFCD09027081 CAS-Nummer: 957062-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...