2-(4-Chlor-3-nitrobenzoyl)benzoesäure

Lieferant: Thermo Scientific

SB00529.1 SB00529.10 SB00529.50
MAYBSB00529.1EA 0 EUR
MAYBSB00529.1 MAYBSB00529.10 MAYBSB00529.50
2-(4-Chlor-3-nitrobenzoyl)benzoesäure
2-(4-Chlor-3-nitrobenzoyl)benzoesäure
Formel: C₁₄H₈ClNO₅
Molecular Weight: 305.67 g/mol
MDL: MFCD00007082
CAS-Nummer: 85-54-1
EINECS: 201-613-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR