2-(4-Chlor-3-nitrobenzoyl)benzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

A13605.18 A13605.30
A13605.18EA 35.3 EUR
A13605.18 A13605.30
2-(4-Chlor-3-nitrobenzoyl)benzoesäure ≥98%
2-(4-Chlor-3-nitrobenzoyl)benzoesäure

Purity: 98% CAS No.: 85-54-1 Molecular Formula: C14H8ClNO5 Formula Weight: 305.68 Melting Point: 198-201° MDL No.: MFCD00007082

Formel: C₁₄H₈ClNO₅
Molecular Weight: 305,67 g/mol
Schmelzpunkt: 198…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007082
CAS-Nummer: 85-54-1
EINECS: 201-613-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR