3-Isothiocyanatopyridin

Lieferant: BIONET RESEARCH

Synonyme: 3-Pyridyl isothiocyanate

6D-048.500 6D-048.1 6D-048.5 6D-048.10
BIOT6D-048.500EA 0 EUR
BIOT6D-048.500 BIOT6D-048.1 BIOT6D-048.5 BIOT6D-048.10
3-Isothiocyanatopyridin
3-Isothiocyanatopyridine
Formel: C₆H₄N₂S
Molecular Weight: 136.18 g/mol
MDL: MFCD00052351
CAS-Nummer: 17452-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR