3-Isothiocyanatopyridin

Lieferant: FLUOROCHEM

Synonyme: 3-Pyridyl isothiocyanate

CFF190.2.5 CFF190.5 CFF190.10 017259-2.5G 017259-5G 017259-10G 017259-1G
CHMPCFF190.2.5EA 268 EUR
CHMPCFF190.2.5 CHMPCFF190.5 CHMPCFF190.10 FLUO017259-2.5G FLUO017259-5G FLUO017259-10G FLUO017259-1G
3-Isothiocyanatopyridin
3-Isothiocyanatopyridine
Formel: C₆H₄N₂S
Molecular Weight: 136.18 g/mol
MDL: MFCD00052351
CAS-Nummer: 17452-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR