3-Isothiocyanatopyridin

Lieferant: Thermo Scientific

Synonyme: 3-Pyridyl isothiocyanate

CC04107DA CC04107EA
MAYBCC04107DAEA 0 EUR
MAYBCC04107DA MAYBCC04107EA
3-Isothiocyanatopyridin
3-Isothiocyanatopyridine
Formel: C₆H₄N₂S
Molecular Weight: 136.18 g/mol
MDL: MFCD00052351
CAS-Nummer: 17452-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR