2-(4-Fluorphenyl)-3-nitropyridin

Lieferant: BIONET RESEARCH

4Y-0709.1 4Y-0709.10 4Y-0709.5 4Y-0709.500
BIOT4Y-0709.1EA 0 EUR
BIOT4Y-0709.1 BIOT4Y-0709.10 BIOT4Y-0709.5 BIOT4Y-0709.500
2-(4-Fluorphenyl)-3-nitropyridin
2-(4-Fluorophenyl)-3-nitropyridine
Formel: C₁₁H₇FN₂O₂
Molecular Weight: 218.19 g/mol
MDL: MFCD06200968
CAS-Nummer: 168267-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR