2-(4-Fluorphenyl)-5-nitropyridin

Lieferant: BIONET RESEARCH

4Y-0704.500 4Y-0704.5 4Y-0704.10 4Y-0704.1
BIOT4Y-0704.500EA 0 EUR
BIOT4Y-0704.500 BIOT4Y-0704.5 BIOT4Y-0704.10 BIOT4Y-0704.1
2-(4-Fluorphenyl)-5-nitropyridin
2-(4-Fluorophenyl)-5-nitropyridine
Formel: C₁₁H₇FN₂O₂
Molecular Weight: 218.19 g/mol
MDL: MFCD06200966
CAS-Nummer: 886361-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR