2-Fluor-5-iodbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 1-Fluoro-4-iodo-2-(trifluoromethyl)benzene

CFF1114. 006245-5G 006245-25G
CHMPCFF1114.EA 0 EUR
CHMPCFF1114. FLUO006245-5G FLUO006245-25G
2-Fluor-5-iodbenzotrifluorid
2-Fluor-5-iodbenzotrifluorid
Formel: C₇H₃F₄I
Molecular Weight: 290 g/mol
MDL: MFCD00236678
CAS-Nummer: 59382-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR