2',4'-Dimethylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4895-1G CA-4895-5G
COBBCA-4895-1GEA 872 EUR
COBBCA-4895-1G COBBCA-4895-5G
2',4'-Dimethylbiphenyl-3-carbonsäure
2',4'-Dimethylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₂
MDL: MFCD12403278
CAS-Nummer: 1170419-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR