2',6'-Dimethylbiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4899-1G
CA-4899-250M
COBBCA-4899-1GEA
1110
EUR
COBBCA-4899-1G
COBBCA-4899-250M
2',6'-Dimethylbiphenyl-3-carbonsäure
2',6'-Dimethylbiphenyl-3-carbonsäure
Formel:
C₁₅H₁₄O₂ |
MDL:
MFCD11895286 CAS-Nummer: 955929-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...