3'-tert-Butylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4874-5G CA-4874-1G CA-4874-250M
COBBCA-4874-5GEA 2510 EUR
COBBCA-4874-5G COBBCA-4874-1G COBBCA-4874-250M
3'-tert-Butylbiphenyl-3-carbonsäure
3'-tert-Butylbiphenyl-3-carbonsäure
Formel: C₁₇H₁₈O₂
MDL: MFCD15143490
CAS-Nummer: 1215206-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR