3'-Methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3'-Methyl[1,1'-biphenyl]-3-carboxylic acid

CA-4832-1G CA-4832-5G
COBBCA-4832-1GEA 50.9 EUR
COBBCA-4832-1G COBBCA-4832-5G
3'-Methylbiphenyl-3-carbonsäure
3'-Methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₂
Molecular Weight: 212.25 g/mol
MDL: MFCD00558892
CAS-Nummer: 158619-46-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR