4'-Methylbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4'-Methyl-[1,1'-biphenyl]-3-carboxylic acid

OT-8004-5G OT-8004-1G
COBBOT-8004-5GEA 249 EUR
COBBOT-8004-5G COBBOT-8004-1G
4'-Methylbiphenyl-3-carbonsäure
4'-Methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
MDL: MFCD00452725
CAS-Nummer: 147404-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR