3-Phenylbenzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

715204-1G 715204-5G
SIAL715204-1GEA 26.6 EUR
SIAL715204-1G SIAL715204-5G
3-Phenylbenzoesäure, Sigma-Aldrich®
3-Phenylbenzoic acid
Formel: C₁₃H₁₀O₂
Molecular Weight: 198,22 g/mol
MDL: MFCD00045846
CAS-Nummer: 716-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR