3'-Methylbiphenyl-4-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
3'-Methyl-[1,1'-biphenyl]-4-carboxylic acid
014239-1G
014239-5G
FLUO014239-1GEA
116
EUR
FLUO014239-1G
FLUO014239-5G
3'-Methylbiphenyl-4-carbonsäure
3'-Methylbiphenyl-4-carbonsäure
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,25 g/mol |
MDL:
MFCD03424590 CAS-Nummer: 5728-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...