4'-Methylbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Methyl-[1,1'-biphenyl]-4-carboxylic acid, 4'-Methyl-4-biphenylcarboxylic acid

010810-1G 010810-5G
FLUO010810-1GEA 45.9 EUR
FLUO010810-1G FLUO010810-5G
4'-Methylbiphenyl-4-carbonsäure
4'-Methylbiphenyl-4-carbonsäure
Formel: C₁₄H₁₂O₂
Molecular Weight: 212.25 g/mol
MDL: MFCD00448778
CAS-Nummer: 720-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR