1-Aminocyclopropancarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Amino-1-cyclopropanecarboxylic acid

M-Clarity™ Qualitätsstufe = MQ200

A3903-100MG A3903-1G A3903-250MG
SIALA3903-100MGEA 104 EUR
SIALA3903-100MG SIALA3903-1G SIALA3903-250MG
1-Aminocyclopropancarbonsäure, Sigma-Aldrich®
1-Aminocyclopropanecarboxylic acid
Formel: C₄H₇NO₂
Molecular Weight: 101,11 g/mol
MDL: MFCD00009944
CAS-Nummer: 22059-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR