Dipraseodymtrisulfat Octahydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Praseodymium(III) sulphate octahydrate
14607.14
14607.22
14607.14EA
57.7
EUR
14607.14
14607.22
Dipraseodymtrisulfat Octahydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Dipraseodymtrisulfat Octahydrat
Purity: 99.9%
CAS No.: 13510-41-3
Molecular Formula: Pr2(SO4)3.8H2O
Formula Weight: 714.12 (570.00anhy)
Physical Form: Crystalline
Appearance: Green
Odour: Odourless
Merck Reference: 14,7712
MDL No.: MFCD00149832
Formel:
O₁₂Pr₂S₃·8H₂O Molecular Weight: 714,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149832 CAS-Nummer: 13510-41-3 EINECS: 233-622-8 Merck Index: 14,7712 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...