4-Vinylbenzoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Styrol-4-carbonsäure
A11107.03
A11107.06
A11107.14
A11107.03EA
94.7
EUR
A11107.03
A11107.06
A11107.14
4-Vinylbenzoesäure ≥98%
4-Vinylbenzoesäure
Purity: 98%
CAS No.: 1075-49-6
Molecular Formula: C9H8O2
Formula Weight: 148.16
Storage Temperature: 2°C to 8°C
Melting Point: ca 142°
Beilstein Reference: 2041130
MDL No.: MFCD00002569
Formel:
C₉H₈O₂ Siedepunkt: 282 °C (1013 hPa) Schmelzpunkt: ∼142 °C Dichte: 1,30 Lagertemperatur: Kühlschrank |
MDL:
MFCD00002569 CAS-Nummer: 1075-49-6 EINECS: 214-053-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...