Allylbenzol 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Phenyl-2-propen, 3-Phenylpropen
102890250
102891000
102895000
ACRO102890250EA
63.2
EUR
ACRO102890250
ACRO102891000
ACRO102895000
Allylbenzol 98%
Allylbenzol
Formel:
(C₆H₅)CH₂CHCH₂ Molecular Weight: 118,18 g/mol Siedepunkt: 156…157 °C (760 mmHg) Schmelzpunkt: ? -40 °C Dichte: 0,89 g/cm³ Flammpunkt: 33 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008651 CAS-Nummer: 300-57-2 EINECS: 206-095-7 UN: 3295 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5095 to 1.5135 (20°C, 589 nm) |
GC | ≥97.5 % |
Appearance of solution | Clear colorless |
Related substances | ≤0.8 % (total impurities) |
pH | 4.0 to 6.0 |
Specific optical rotation | +33.7° to +36.0° (20°C, 589 nm) (on dried substance) |
Titration with HClO4 | 99.0 to 101.0 % (on dried substance) |
Sulfates | ≤200 ppm |
Chlorides | ≤200 ppm |
Ammonium (NH4) | ≤0.1 % |
Iron (Fe) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...