3,4-Dibromtoluol

Lieferant: FLUOROCHEM

Synonyme: 1,2-Dibromo-4-methylbenzene

018506-1G 018506-5G
FLUO018506-1GEA 0 EUR
FLUO018506-1G FLUO018506-5G
3,4-Dibromtoluol
3,4-Dibromtoluol
Formel: C₇H₆Br₂
MDL: MFCD00079744
CAS-Nummer: 60956-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR