α,3,5-Tribromtoluol ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3,5-Dibromobenzyl bromide, 1,3-Dibromo-5-(bromomethyl)benzene, alpha,3,5-Tribromotoluene
A12726.03
A12726.06
A12726.14
A12726.03EA
51.6
EUR
A12726.03
A12726.06
A12726.14
α,3,5-Tribromtoluol ≥99%
α,3,5-Tribromtoluol
Purity: 99%
CAS No.: 56908-88-4
Molecular Formula: C7H5Br3
Formula Weight: 328.84
Melting Point: 93-95°
Beilstein Reference: 3237827
UN No.: 1759
MDL No.: MFCD00052415
Formel:
C₇H₅Br₃ Molecular Weight: 328,83 g/mol Siedepunkt: 173 °C (19 mmHg) Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052415 CAS-Nummer: 56908-88-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...