1,4,8,11-Tetraazacyclotetradecan 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: CYCLAM

269330010 269330050 269330250
ACRO269330010EA 63.4 EUR
ACRO269330010 ACRO269330050 ACRO269330250
1,4,8,11-Tetraazacyclotetradecan 98%
1,4,8,11-Tetraazacyclotetradecane
Formel: C₁₀H₂₄N₄
Molecular Weight: 200,33 g/mol
Schmelzpunkt: 185…188 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005105
CAS-Nummer: 295-37-4
EINECS: 206-039-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or needles
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR