4-Methyl-2-pentensäure ≥98%

Lieferant: Thermo Scientific
Gefahr

B22639.09 B22639.18
B22639.09EA 34.2 EUR
B22639.09 B22639.18
4-Methyl-2-pentensäure ≥98%
4-Methyl-2-pentenoic acid

Purity: 98+% CAS No.: 10321-71-8 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 100°(212°F) Refractive Index: 1.4485 Odour: Unpleasant Boiling Point: 212° UN No.: 3265 MDL No.: MFCD00043804

Formel: C₆H₁₀O₂
Molecular Weight: 114,14 g/mol
Siedepunkt: 212 °C
Dichte: 0,959
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00043804
CAS-Nummer: 10321-71-8
EINECS: 233-706-4
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR