1-Benzyloxy-4-brombenzol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Benzyloxybromobenzene, 1-Bromo-4-(phenylmethoxy)benzene
B24923.03
B24923.06
B24923.14
B24923.03EA
20.2
EUR
B24923.03
B24923.06
B24923.14
1-Benzyloxy-4-brombenzol ≥97%
1-Benzyloxy-4-brombenzol
Purity: 97%
CAS No.: 6793-92-6
Molecular Formula: C13H11BrO
Formula Weight: 263.14
Melting Point: 61-63°
Boiling Point: 166-167°/3mm
MDL No.: MFCD00028016
Formel:
C₁₃H₁₁BrO Molecular Weight: 263.13 Siedepunkt: 166…167 °C (3 mmHg) Schmelzpunkt: 61…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00028016 CAS-Nummer: 6793-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...